1-(Difluoromethoxy)-4-nitrobenzene 97%

Supplier: Ambeed
Warning

A179728-100G A179728-1G A179728-25G
77111-368EA 106.09 USD
77111-368 77111-370 77111-372
1-(Difluoromethoxy)-4-nitrobenzene 97%
1-(Difluoromethoxy)-4-nitrobenzene

Precaución: for Research Use Only

Formula: C₇H₅F₂NO₃
MDL Number: MFCD00085009
CAS Number: 1544-86-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR