3-(Morpholino)phenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: (3-Morpholinophenyl)boronic acid

A179301-10G A179301-1G A179301-250MG
77111-356EA 159.64 USD
77111-356 77111-358 77567-644
3-(Morpholino)phenylboronic acid 97%
3-(Morpholino)phenylboronic acid

Precaución: For research use only.

Formula: C₁₀H₁₄BNO₃
MW: 207.0341 g/mol
MDL Number: MFCD03412095
CAS Number: 863377-22-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR