4-Amino-2-methylpyrimidine-5-carbonitrile 97%

Supplier: Ambeed
Warning

A178311-10G A178311-1G A178311-250MG A178311-5G
77111-312EA 98.67 USD
77111-312 77111-314 77111-316 77111-318
4-Amino-2-methylpyrimidine-5-carbonitrile 97%
4-Amino-2-methylpyrimidine-5-carbonitrile

Precaución: for Research Use Only

Formula: C₆H₆N₄
MDL Number: MFCD00084875
CAS Number: 698-29-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR