Methyl-2-aminothiophene-3-carboxylate 97%
Supplier: Ambeed
A178298-100G
A178298-10G
A178298-1G
A178298-25G
A178298-500G
A178298-5G
77111-300EA
55.62
USD
77111-300
77111-302
77111-304
77111-306
77111-308
77111-310
Methyl-2-aminothiophene-3-carboxylate 97%
Methyl-2-aminothiophene-3-carboxylate
Precaución: for Research Use Only
Formula:
C₆H₇NO₂S |
MDL Number:
MFCD00159547 CAS Number: 4651-81-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...