Captopril 99%

Supplier: Ambeed
Danger

A177871-1G A177871-25G A177871-5G A177871-100G
77111-278EA 15.45 USD
77111-278 77111-280 77111-282 77685-400
Captopril 99%
Captopril

Precaución: for Research Use Only

Formula: C₉H₁₅NO₃S
MDL Number: MFCD00168073
CAS Number: 62571-86-2
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR