2,3,4-Trifluorotoluene 99%

Supplier: Ambeed
Danger

A177763-1G A177763-25G A177763-5G
77111-272EA 9.55 USD
77111-272 77111-274 77111-276
2,3,4-Trifluorotoluene 99%
2,3,4-Trifluorotoluene

Precaución: for Research Use Only

Formula: C₇H₅F₃
MW: 146.1098 g/mol
MDL Number: MFCD00075194
CAS Number: 193533-92-5
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR