3-Bromo-2-fluoro-4-iodopyridine 95%

Supplier: Ambeed
Warning

A177285-250MG A177285-5G A177285-1G
77111-264EA 15.45 USD
77111-264 77111-266 77593-130
3-Bromo-2-fluoro-4-iodopyridine 95%
3-Bromo-2-fluoro-4-iodopyridine

Precaución: for Research Use Only

Formula: C₅H₂BrFIN
MDL Number: MFCD03095311
CAS Number: 884494-52-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR