L-Chicoric acid 98%

Supplier: Ambeed
Warning

A176803-10MG A176803-1MG A176803-5MG
77111-240EA 48.42 USD
77111-240 77111-242 77111-244
L-Chicoric acid 98%
L-Chicoric acid

Precaución: for Research Use Only

Formula: C₂₂H₁₈O₁₂
MDL Number: MFCD22683653
CAS Number: 70831-56-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR