α-Acetyl-γ-butyrolactone 95%

Supplier: Ambeed
Warning

Synonyms: alpha-Acetyl-gamma-hydroxybutyric acid Lactone, α-Acetyl-γ-hydroxybutyric acid Lactone, 3-Acetyldihydrofuran-2(3H)-one, 2-Acetyl-γ-butyrolactone, 2-Acetylbutyrolactone, alpha-Acetyl-gamma-butyrolactone, α-Acetylbutyrolactone, 3-Acetyl-4,5-dihydro-2(3H)-furanone

A176739-1000G A176739-100G A176739-500G A176739-25G
77111-234EA 50.47 USD
77111-234 77111-236 77111-238 77615-488
α-Acetyl-γ-butyrolactone 95%
α-Acetyl-γ-butyrolactone

Precaución: for Research Use Only

Formula: C₆H₈O₃
MW: 128.13 g/mol
MDL Number: MFCD00005394
CAS Number: 517-23-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR