3-Chloro-5-fluoropyridine 97%

Supplier: Ambeed
Danger

A176230-10G A176230-250MG A176230-25G A176230-5G
77111-202EA 27.81 USD
77111-202 77111-204 77111-206 77111-208
3-Chloro-5-fluoropyridine 97%
3-Chloro-5-fluoropyridine

Precaución: for Research Use Only

Formula: C₅H₃ClFN
MDL Number: MFCD05663707
CAS Number: 514797-99-0
UN: 1325
ADR: 4.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR