3-Bromo-4-chlorobenzoic acid 97%

Supplier: Ambeed
Warning

A175497-100G A175497-10G A175497-25G A175497-5G A175497-500G
77111-194EA 40.17 USD
77111-194 77111-196 77111-198 77111-200 77685-393
3-Bromo-4-chlorobenzoic acid 97%
3-Bromo-4-chlorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄BrClO₂
MDL Number: MFCD00079706
CAS Number: 42860-10-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR