Ethyl-2-amino-4-methylthiazole-5-carboxylate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ethyl 2-amino-4-methylthiazole-5-carboxylate
A175434-100G
A175434-10G
A175434-1G
A175434-25G
A175434-5G
77111-184EA
94.43
USD
77111-184
77111-186
77111-188
77111-190
77111-192
Ethyl-2-amino-4-methylthiazole-5-carboxylate 97%
Ethyl 2-amino-4-methylthiazole-5-carboxylate
Precaución: for Research Use Only
Formula:
C₇H₁₀N₂O₂S MW: 186.22 g/mol |
MDL Number:
MFCD00123414 CAS Number: 7210-76-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...