2,3,5,6-Tetrafluorothiophenol 98%

Supplier: Ambeed
Danger

Synonyms: 1-Mercapto-2,3,5,6-tetrafluorobenzene, 2,3,5,6-Tetrafluorobenzenethiol, 2,3,5,6-Tetrafluorobenzene-1-thiol, 2,3,5,6-tetrafluorothiophenol

A175267-1G A175267-25G A175267-5G
77111-176EA 18.04 USD
77111-176 77111-178 77111-180
2,3,5,6-Tetrafluorothiophenol 98%
2,3,5,6-Tetrafluorothiophenol

Precaución: for Research Use Only

Formula: C₆H₂F₄S
MW: 182.14 g/mol
MDL Number: MFCD00004829
CAS Number: 769-40-4
UN: 1992
ADR: 3,6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR