1,2-Diamino-3,4-difluorobenzene 98%

Supplier: Ambeed
Warning

Synonyms: 3,4-Difluoro-1,2-phenylenediamine, 3,4-Difluorobenzene-1,2-diamine, 3,4-Difluoro-o-phenylenediamine

A174983-100MG A174983-250MG
77111-172EA 5.31 USD
77111-172 77111-174
1,2-Diamino-3,4-difluorobenzene 98%
1,2-Diamino-3,4-difluorobenzene

Precaución: for Research Use Only

Formula: C₆H₆F₂N₂
MW: 144.12 g/mol
MDL Number: MFCD00153128
CAS Number: 153505-39-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR