2-Chloro-4,5-difluorobenzoic acid 97%

Supplier: Ambeed
Warning

A174727-100G A174727-1G A174727-25G A174727-5G
77111-148EA 163.38 USD
77111-148 77111-150 77111-152 77111-154
2-Chloro-4,5-difluorobenzoic acid 97%
2-Chloro-4,5-difluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃ClF₂O₂
MW: 192.55 g/mol
MDL Number: MFCD00077479
CAS Number: 110877-64-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR