1-Bromo-2,3,4-trimethoxybenzene 97%

Supplier: Ambeed
Warning

A173775-1G A173775-250MG A173775-5G
77111-120EA 13.39 USD
77111-120 77111-122 77111-124
1-Bromo-2,3,4-trimethoxybenzene 97%
1-Bromo-2,3,4-trimethoxybenzene

Precaución: for Research Use Only

Formula: C₉H₁₁BrO₃
MDL Number: MFCD01318955
CAS Number: 10385-36-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR