6-Methyl-3(2H)-pyridazinone 97%

Supplier: Ambeed
Warning

A173363-100G A173363-10G A173363-1G A173363-25G A173363-5G
77111-094EA 57.67 USD
77111-094 77111-096 77111-098 77111-100 77111-102
6-Methyl-3(2H)-pyridazinone 97%
6-Methyl-3(2H)-pyridazinone

Precaución: for Research Use Only

Formula: C₅H₆N₂O
MDL Number: MFCD00039720
CAS Number: 13327-27-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR