Butylmalonic acid 98%

Supplier: Ambeed
Warning

Synonyms: Butyl-1,3-propanedioic acid, 2-Butylmalonic acid

A172808-100G A172808-5G
77111-072EA 729.9 USD
77111-072 77111-074
Butylmalonic acid 98%
Butylmalonic acid

Precaución: for Research Use Only

Formula: C₇H₁₂O₄
MW: 160.17 g/mol
MDL Number: MFCD00002673
CAS Number: 534-59-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR