4-(Difluoromethyl)picolinic acid 95%

Supplier: Ambeed
Warning

A172531-100MG A172531-1G
77111-068EA 158.08 USD
77111-068 77111-070
4-(Difluoromethyl)picolinic acid 95%
4-(Difluoromethyl)picolinic acid

Precaución: for Research Use Only

Formula: C₇H₅F₂NO₂
MW: 173.12 g/mol
MDL Number: MFCD18256210
CAS Number: 1256804-49-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR