N'-(7-Fluoropyrrolo[1,2-a]quinoxalin-4-yl)pyrazine-2-carbohydrazide ≥98%

Supplier: Ambeed
Warning

Synonyms: SC144

A172019-10MG A172019-1MG A172019-50MG A172019-5MG
77111-052EA 59.74 USD
77111-052 77111-054 77111-056 77111-058
N'-(7-Fluoropyrrolo[1,2-a]quinoxalin-4-yl)pyrazine-2-carbohydrazide ≥98%
N'-(7-Fluoropyrrolo[1,2-a]quinoxalin-4-yl)pyrazine-2-carbohydrazide

Precaución: for Research Use Only

Formula: C₁₆H₁₁FN₆O
MDL Number: MFCD25976763
CAS Number: 895158-95-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR