4-Amino-3,5-dibromobenzonitrile 95%
Supplier: Ambeed
|
Warning
|
A171356-100G
A171356-10G
A171356-1G
A171356-250MG
A171356-25G
A171356-5G
77111-018EA
262.05
USD
77111-018
77111-020
77111-022
77111-024
77111-026
77111-028
4-Amino-3,5-dibromobenzonitrile 95%
4-Amino-3,5-dibromobenzonitrile
Precaución: for Research Use Only
Formula:
C₇H₄Br₂N₂ MW: 275.9300 g/mol |
MDL Number:
MFCD03094688 CAS Number: 58633-04-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...