2,5-Dichloropyrimidine 98%

Supplier: Ambeed
Warning

A170857-100G A170857-10G A170857-25G A170857-5G A170857-1G
77110-996EA 154.9 USD
77110-996 77110-998 77111-000 77111-002 77593-064
2,5-Dichloropyrimidine 98%
2,5-Dichloropyrimidine

Precaución: for Research Use Only

Formula: C₄H₂Cl₂N₂
MW: 148.97 g/mol
MDL Number: MFCD00673131
CAS Number: 22536-67-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR