Cyclohexylbenzene 97%

Supplier: Ambeed
Danger

Synonyms: Phenylcyclohexane

A170850-10G A170850-100G
77110-994EA 5.31 USD
77110-994 77685-378
Cyclohexylbenzene 97%
Cyclohexylbenzene

Precaución: for Research Use Only

Formula: C₁₂H₁₆
MW: 160.26 g/mol
MDL Number: MFCD00001451
CAS Number: 827-52-1
UN: 3082
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR