1-Bromonaphthalene 97%

Supplier: Ambeed
Warning

Synonyms: α-Bromonaphthalene

A170548-100G A170548-10G A170548-25G A170548-500G
77110-970EA 12.74 USD
77110-970 77110-972 77110-974 77550-098
1-Bromonaphthalene 97%
1-Bromonaphthalene

Precaución: for Research Use Only

Formula: C₁₀H₇Br
MW: 207.07 g/mol
MDL Number: MFCD00003868
CAS Number: 90-11-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR