Benzyl-4-bromophenyl ketone 97%

Supplier: Ambeed
Warning

A170334-10G A170334-1G A170334-250MG A170334-25G A170334-5G
77110-960EA 155.53 USD
77110-960 77110-962 77110-964 77110-966 77110-968
Benzyl-4-bromophenyl ketone 97%
Benzyl 4-bromophenyl ketone

Precaución: for Research Use Only

Formula: C₁₄H₁₁BrO
MDL Number: MFCD00016331
CAS Number: 2001-29-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR