2-Bromo-5-chlorobenzonitrile 98%

Supplier: Ambeed
Warning

A170262-100G A170262-5G
77110-956EA 184.6 USD
77110-956 77110-958
2-Bromo-5-chlorobenzonitrile 98%
2-Bromo-5-chlorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃BrClN
MW: 216.46 g/mol
MDL Number: MFCD08445660
CAS Number: 57381-37-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR