2,4-Diamino-6-piperidinopyrimidine 98%

Supplier: Ambeed
Warning

A169864-100MG A169864-1G A169864-250MG A169864-5G
77110-948EA 26.53 USD
77110-948 77110-950 77110-952 77110-954
2,4-Diamino-6-piperidinopyrimidine 98%
2,4-Diamino-6-piperidinopyrimidine

Precaución: for Research Use Only

Formula: C₉H₁₅N₅
MW: 193.25 g/mol
MDL Number: MFCD04973589
CAS Number: 24867-26-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR