Creatine phosphate disodium salt tetrahydrate 96%

Supplier: Ambeed
Warning

Synonyms: Sodium creatine phosphate dibasic tetrahydrate, Sodium N-carbamimidoyl-N-methylglycinate dihydrogen phosphate tetrahydrate, Phosphocreatine disodium salt tetrahydrate

A169586-100G A169586-1G A169586-25G A169586-5G A169586-10G A169586-500G
77110-940EA 200.85 USD
77110-940 77110-942 77110-944 77110-946 77685-373 77685-374
Creatine phosphate disodium salt tetrahydrate 96%
Creatine phosphate disodium salt tetrahydrate

Precaución: for Research Use Only

Formula: C₄H₈N₃Na₂O₅P·4H₂O
MW: 327.14 g/mol (255.07 anhy)
MDL Number: MFCD00150192
CAS Number: 71519-72-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR