TC-E 5003 95%

Supplier: Ambeed
Warning

Synonyms: N,N'-(sulphonyldi-4,1-phenylene)bis(2-chloroacetamide, N,N'-(sulphonylbis(4,1-phenylene))bis(2-chloroacetamide)

A169549-10MG A169549-1MG A169549-25MG A169549-50MG A169549-5MG
77110-930EA 44.56 USD
77110-930 77110-932 77110-934 77110-936 77110-938
TC-E 5003 95%
TC-E 5003

Precaución: for Research Use Only

Formula: C₁₆H₁₄Cl₂N₂O₄S
MW: 401.26 g/mol
MDL Number: MFCD00028183
CAS Number: 17328-16-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR