3-(Trimethylsilyl)-2-propyn-1-ol 97%

Supplier: Ambeed
Warning

Synonyms: 3-(Trimethylsilyl)propargyl alcohol

A167688-100G A167688-10G A167688-1G A167688-25G A167688-5G
77110-898EA 135.96 USD
77110-898 77110-900 77110-902 77110-904 77110-906
3-(Trimethylsilyl)-2-propyn-1-ol 97%
3-(Trimethylsilyl)-2-propyn-1-ol

Precaución: for Research Use Only

Formula: C₆H₁₂OSi
MDL Number: MFCD00002913
CAS Number: 5272-36-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR