2-Amino-6-ethylpyridine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
6-Ethylpyridin-2-amine, 6-Ethyl-2-pyridylamine
A167174-10G
A167174-1G
A167174-25G
A167174-5G
77110-874EA
103.97
USD
77110-874
77110-876
77110-878
77110-880
2-Amino-6-ethylpyridine 97%
2-Amino-6-ethylpyridine
Precaución: for Research Use Only
Formula:
C₇H₁₀N₂ MW: 122.17 g/mol |
MDL Number:
MFCD00218265 CAS Number: 21717-29-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...