DL-2,3-Diaminopropionic acid hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A166884-100G
A166884-10G
A166884-1G
A166884-25G
A166884-5G
77110-860EA
294.58
USD
77110-860
77110-862
77110-864
77110-866
77110-868
DL-2,3-Diaminopropionic acid hydrochloride 97%
DL-2,3-Diaminopropionic acid hydrochloride
Precaución: for Research Use Only
Formula:
C₃H₈N₂O₂ |
MDL Number:
MFCD00012884 CAS Number: 54897-59-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...