Naringin 98%

Supplier: Ambeed
Warning

Synonyms: (S)-7-(((2S,3R,4S,5S,6R)-4,5-Dihydroxy-6-(hydroxymethyl)-3-(((2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2-yl)oxy)-5-hydroxy-2-(4-hydroxyphenyl)chroman-4-one, 4',5,7-Trihydroxyflavanone 7-rhamnoglucoside, Naringenin 7-rhamnoglucoside

A165939-10G A165939-25G A165939-5G A165939-100G
77110-846EA 12.37 USD
77110-846 77110-848 77110-850 77567-416
Naringin 98%
Naringin

Precaución: For research use only.

Formula: C₂₇H₃₂O₁₄
MDL Number: MFCD00148888
CAS Number: 10236-47-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR