3,4-Difluoro-o-toluic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3,4-Difluoro-2-methylbenzoic acid

A165825-100G A165825-10G A165825-1G A165825-250MG A165825-25G A165825-5G
77110-834EA 336.81 USD
77110-834 77110-836 77110-838 77110-840 77110-842 77110-844
3,4-Difluoro-o-toluic acid 97%
3,4-Difluoro-o-toluic acid

Precaución: for Research Use Only

Formula: C₈H₆F₂O₂
MW: 172.13 g/mol
MDL Number: MFCD07777153
CAS Number: 157652-31-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR