4-Chloro-3-nitrobenzonitrile 97%
Supplier: Ambeed
|
Warning
|
A165676-100G
A165676-10G
A165676-1G
A165676-25G
A165676-500G
A165676-5G
77110-822EA
77.45
USD
77110-822
77110-824
77110-826
77110-828
77110-830
77110-832
4-Chloro-3-nitrobenzonitrile 97%
4-Chloro-3-nitrobenzonitrile
Precaución: for Research Use Only
Formula:
C₇H₃ClN₂O₂ MW: 182.57 g/mol |
MDL Number:
MFCD00016987 CAS Number: 939-80-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...