Ethyl-3-amino-1H-pyrazole-4-carboxylate 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Amino-4-carbethoxypyrazole, Ethyl 3-amino-1H-pyrazole-4-carboxylate
A164006-100G
A164006-10G
A164006-1G
A164006-25G
A164006-500G
A164006-5G
77110-774EA
51.99
USD
77110-774
77110-776
77110-778
77110-780
77110-782
77110-784
Ethyl-3-amino-1H-pyrazole-4-carboxylate 95%
Ethyl 3-amino-1H-pyrazole-4-carboxylate
Precaución: for Research Use Only
Formula:
C₆H₉N₃O₂ MW: 155.16 g/mol |
MDL Number:
MFCD00005238 CAS Number: 6994-25-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...