(4-(Difluoromethoxy)-3,5-difluorophenyl)boronic acid 98%

Supplier: Ambeed
Warning

A163902-1G A163902-250MG A163902-5G
77110-768EA 164.44 USD
77110-768 77110-770 77110-772
(4-(Difluoromethoxy)-3,5-difluorophenyl)boronic acid 98%
(4-(Difluoromethoxy)-3,5-difluorophenyl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₅BF₄O₃
MW: 223.92 g/mol
MDL Number: MFCD08272029
CAS Number: 915401-97-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR