5-Chloro-2-hydroxypyrazine 98%
Supplier: Ambeed
|
Warning
|
A162367-1G
A162367-5G
77110-748EA
35.01
USD
77110-748
77110-750
5-Chloro-2-hydroxypyrazine 98%
5-Chloro-2-hydroxypyrazine
Precaución: for Research Use Only
Formula:
C4H3ClN2O MW: 130.5300 g/mol |
MDL Number:
MFCD11109884 CAS Number: 89180-45-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...