4,6-Dichloropicolinaldehyde 95%

Supplier: Ambeed
Warning

A162002-100MG A162002-250MG A162002-1G
77110-736EA 94.43 USD
77110-736 77110-738 77550-028
4,6-Dichloropicolinaldehyde 95%
4,6-Dichloropicolinaldehyde

Precaución: for Research Use Only

Formula: C₆H₃Cl₂NO
MW: 176 g/mol
MDL Number: MFCD10698614
CAS Number: 132683-62-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR