4-Bromo-2-chloro-5-fluoropyridine 97%

Supplier: Ambeed
Warning

A161528-10G A161528-1G A161528-250MG A161528-5G
77110-728EA 795.68 USD
77110-728 77110-730 77110-732 77110-734
4-Bromo-2-chloro-5-fluoropyridine 97%
4-Bromo-2-chloro-5-fluoropyridine

Precaución: for Research Use Only

Formula: C₅H₂BrClFN
MW: 210.431 g/mol
MDL Number: MFCD06659500
CAS Number: 884495-10-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR