4-Amino-5-bromo-3-methylpyridine 97%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-5-methylpyridin-4-amine

A161498-10G A161498-1G A161498-250MG A161498-25G A161498-5G
77110-718EA 180.36 USD
77110-718 77110-720 77110-722 77110-724 77110-726
4-Amino-5-bromo-3-methylpyridine 97%
4-Amino-5-bromo-3-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₇BrN₂
MW: 187.0372 g/mol
MDL Number: MFCD07437841
CAS Number: 97944-43-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR