NSC 405020 98%

Supplier: Ambeed
Warning

Synonyms: 3,4-Dichloro-N-(pentan-2-yl)benzamide

A160858-10MG A160858-1MG A160858-5MG
77110-694EA 59.75 USD
77110-694 77110-696 77110-698
NSC 405020 98%
NSC 405020

Precaución: for Research Use Only

Formula: C₁₂H₁₅Cl₂NO
MDL Number: MFCD02006898
CAS Number: 7497-07-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR