4,6-Dichloro-2-methyl-5-nitropyrimidine 97%
Supplier: Ambeed
|
Warning
|
A160657-100G
A160657-10G
A160657-1G
A160657-250MG
A160657-25G
A160657-5G
77110-674EA
414.82
USD
77110-674
77110-676
77110-678
77110-680
77110-682
77110-684
4,6-Dichloro-2-methyl-5-nitropyrimidine 97%
4,6-Dichloro-2-methyl-5-nitropyrimidine
Precaución: for Research Use Only
Formula:
C₅H₃Cl₂N₃O₂ MW: 208.0000 g/mol |
MDL Number:
MFCD00060527 CAS Number: 13162-43-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...