2'-Bromo-5'-phenyl-1,1':3',1''-terphenyl 98%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-1,3,5-triphenylbenzene, 2'-Bromo-5'-phenyl-m-terphenyl

A160624-100MG A160624-1G A160624-250MG A160624-25G A160624-5G
77110-664EA 6.37 USD
77110-664 77110-666 77110-668 77110-670 77110-672
2'-Bromo-5'-phenyl-1,1':3',1''-terphenyl 98%
2'-Bromo-5'-phenyl-1,1':3',1''-terphenyl

Precaución: for Research Use Only

Formula: C₂₄H₁₇Br
MW: 385.30 g/mol
MDL Number: MFCD00092320
CAS Number: 10368-73-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR