4-Fluoro-5-methoxy-2-methyl-1H-indole 97%

Supplier: Ambeed
Warning

A160613-1G A160613-250MG
77110-660EA 35.01 USD
77110-660 77110-662
4-Fluoro-5-methoxy-2-methyl-1H-indole 97%
4-Fluoro-5-methoxy-2-methyl-1H-indole

Precaución: for Research Use Only

Formula: C₁₀H₁₀FNO
MDL Number: MFCD09264512
CAS Number: 288385-93-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR