(3-Chlorophenyl)hydrazine hydrochloride 98%

Supplier: Ambeed
Warning

A160582-1000G A160582-100G A160582-10G A160582-500G
77110-652EA 223.51 USD
77110-652 77110-654 77110-656 77110-658
(3-Chlorophenyl)hydrazine hydrochloride 98%
(3-Chlorophenyl)hydrazine hydrochloride

Precaución: for Research Use Only

Formula: C₆H₈Cl₂N₂
MDL Number: MFCD00012935
CAS Number: 2312-23-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR