[Ru(bpz)3][PF6]2 98%

Supplier: Ambeed
Warning

A160343-100MG A160343-1G A160343-250MG A160343-50MG
77110-640EA 167.63 USD
77110-640 77110-642 77110-644 77110-646
[Ru(bpz)3][PF6]2 98%
[Ru(bpz)3][PF6]2

Precaución: for Research Use Only

Formula: C₂₄H₁₈F₁₂N₁₂P₂Ru
MW: 865.47 g/mol
MDL Number: MFCD27952557
CAS Number: 80907-56-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR