(S)-4-Amino-2-(benzyloxycarbonylamino)butyric acid 98%

Supplier: Ambeed
Warning

Synonyms: (S)-4-Amino-2-(((benzyloxy)carbonyl)amino)butanoic acid, Z-Dab-OH, Cbz-α-Dab-OH

A160322-5G A160322-25G
77110-632EA 67.98 USD
77110-632 77685-312
(S)-4-Amino-2-(benzyloxycarbonylamino)butyric acid 98%
(S)-4-Amino-2-(benzyloxycarbonylamino)butyric acid

Precaución: for Research Use Only

Formula: C₁₂H₁₆N₂O₄
MDL Number: MFCD00077904
CAS Number: 62234-40-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR