2,5-Diamino-1,4-benzenedithiol dihydrochloride 97% (by HPLC)

Supplier: Ambeed
Danger

A159905-1G A159905-250MG A159905-25G A159905-5G
77110-618EA 17.52 USD
77110-618 77110-620 77110-622 77110-624
2,5-Diamino-1,4-benzenedithiol dihydrochloride 97% (by HPLC)
2,5-Diamino-1,4-benzenedithiol dihydrochloride

Precaución: for Research Use Only

Formula: C₆H₈N₂S₂
MDL Number: MFCD00142835
CAS Number: 75464-52-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR