5-Bromo-1,3-benzodioxole 98%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-1,2-(methylenedioxy)benzene, 5-Bromobenzo[d][1,3]dioxole

A159811-100G A159811-10G A159811-1G A159811-25G A159811-500G A159811-5G A159811-1000G
77110-604EA 28.65 USD
77110-604 77110-606 77110-608 77110-610 77110-612 77110-614 77615-304
5-Bromo-1,3-benzodioxole 98%
5-Bromo-1,3-benzodioxole

Precaución: for Research Use Only

Formula: C₇H₅BrO₂
MW: 201.02 g/mol
MDL Number: MFCD00005821
CAS Number: 2635-13-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR